ω,4-Dibromoacetophenone for synthesis, Sigma-Aldrich®

Danger

Synonyms: 2-Bromo-1-(4-bromophenyl)-1-ethanoneGlycogen Synthase Kinase-3β Inhibitor VII2,4'-DibromoacetophenoneGSK-3β Inhibitor VIITPK I Inhibitor4-Bromophenacyl bromideTau Protein Kinase I Inhibitorα,4-Dibromoacetophenone

M-Clarity™ quality level = MQ200

8.22019.0025 8.22019.0025
EM8.22019.0025CS 8525.04 USD
EM8.22019.0025 CA8.22019.0025
ω,4-Dibromoacetophenone for synthesis, Sigma-Aldrich®
ω,4-Dibromoacetophenone
Formula: C₈H₆Br₂O
MW: 277.94 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 107…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000200
CAS Number: 99-73-0
EINECS: 202-783-6
UN: 1759
ADR: 8,III
Merck Index: 13,01412

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 107 - 110 °C
Molar Mass 277.94 g/mol
Boiling Point 1415 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR