Tetraoctyltin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Tetra-n-octyltinTetraoctylstannane

M-Clarity™ quality level = MQ200

8.21898.0100 8.21898.0100
EM8.21898.0100CS 2314 USD
CA8.21898.0100 EM8.21898.0100
Tetraoctyltin for synthesis, Sigma-Aldrich®
Tetraoctyltin
Formula: C₃₂H₆₈Sn
MW: 571.6 g/mol
Boiling Pt: 268 °C (13 hPa)
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
CAS Number: 3590-84-9
EINECS: 222-733-7
UN: 2788
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Molar Mass 571.57 g/mol
Boiling Point 268 °C (13 hPa)
Flash Point 180 °C
Refractive Index 1.4681 (20 °C, 589 nm)
Density 0.98 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR