Phenylmercury hydroxide for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.21819.0050 8.21819.0050
EM8.21819.0050CS 12175.75 USD
CA8.21819.0050 EM8.21819.0050
Phenylmercury hydroxide for synthesis, Sigma-Aldrich®
Phenylmercury hydroxide
Formula: C₆H₆HgO
CAS Number: 100-57-2
EINECS: 202-866-7
UN: 1894
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 50 g/l (75 °C)
Molar Mass 294.7 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR