2-Bromohexane mixture of isomers for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21707.0025 8.21707.0025
EM8.21707.0025CS 18953 USD
EM8.21707.0025 CA8.21707.0025
2-Bromohexane mixture of isomers for synthesis, Sigma-Aldrich®
2-Bromohexane
Formula: H₃CBrCH(CH₂)₃CH₃
MW: 165.07 g/mol
Boiling Pt: 142…144 °C (1013 hPa)
Density: 1.166 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039183
CAS Number: 3377-86-4
EINECS: 222-173-3
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 165.08 g/mol
Boiling Point 142 - 144 °C (1013 hPa)
Vapor Pressure 9.7 hPa (25 °C)
Flash Point 34 °C
Refractive Index 1.4432 (25 °C, 589 nm)
Density 1.166 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR