Tributyltin acetate for synthesis, Sigma-Aldrich®

Danger

Synonyms: Tri-n-butyltin acetate

M-Clarity™ quality level = MQ200

8.21650.0050
EM8.21650.0050CS 2220.88 USD
EM8.21650.0050
Tributyltin acetate for synthesis, Sigma-Aldrich®
Tributyltin acetate
Formula: C₁₄H₃₀O₂Sn
MW: 349.1 g/mol
MDL Number: MFCD00015228
CAS Number: 56-36-0
EINECS: 200-269-6
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 0.065 g/l (20 °C)
Melting Point 79 - 82 °C
Molar Mass 349.07 g/mol
Vapor Pressure 0.001 hPa (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR