Triphenylantimony for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Triphenylstibine
M-Clarity™ quality level = MQ200
8.21199.0100
8.21199.0100
EM8.21199.0100CS
2126
USD
CA8.21199.0100
EM8.21199.0100
Triphenylantimony for synthesis, Sigma-Aldrich®
Triphenylantimony
Formula:
C₁₈H₁₅Sb MW: 353.08 g/mol Boiling Pt: 377 °C (1013 hPa) Melting Pt: 52…54 °C Density: 1.5 g/cm³ (12 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003000 CAS Number: 603-36-1 EINECS: 210-037-6 UN: 2811 ADR: 6.1,III |
Specification Test Results
Solubility |
|
Melting Point | 52 - 54 °C |
Molar Mass | 353.07 g/mol |
Bulk Density | 1490 kg/m3 |
Boiling Point | 377 °C (1013 hPa) |
Flash Point | 113 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...