Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tributylphosphine oxide for synthesis

Supplier: MilliporeSigma

Warning

8.21145.0025 8.21145.0025 8.21145.0005 8.21145.0005
EM8.21145.0025CS 10286.3 USD InStock
EM8.21145.0025 CA8.21145.0025 CA8.21145.0005 EM8.21145.0005
Tributylphosphine oxide for synthesis
Tributylphosphine oxide


Formula: C₁₂H₂₇OP
MW: 218,32 g/mol
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 62 - 63 °C
MDL Number: MFCD00002082
CAS Number: 814-29-9
EINECS: 212-394-3
Order Now
Specification Test Results
Solubility 55.7 g/l (25 °C)
Melting Point 68 - 71 °C
Molar Mass 218.32 g/mol
Boiling Point 150 °C (2 hPa)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty