2-Oxobutyric acid for synthesis, Sigma-Aldrich®

Danger

Synonyms: 2-Ketobutyric acidα-Ketobutyric acid2-Oxobutanoic acid

M-Clarity™ quality level = MQ200

8.20938.0005 8.20938.0005
EM8.20938.0005CS 6739.73 USD
EM8.20938.0005 CA8.20938.0005
2-Oxobutyric acid for synthesis, Sigma-Aldrich®
2-Oxobutyric acid
Formula: C₄H₆O₃
MW: 102.09 g/mol
MDL Number: MFCD00004164
CAS Number: 600-18-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Solubility 100 g/l (20 °C)
Melting Point 31 - 34 °C
Molar Mass 102.08 g/mol
Boiling Point 68 °C (20 hPa)
Flash Point 81 °C
pH 1.1 (100 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR