Methylcyclopentane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20789.0005 8.20789.0250 8.20789.0250 8.20789.0005
EM8.20789.0005CS 2339 USD
EM8.20789.0005 CA8.20789.0250 EM8.20789.0250 CA8.20789.0005
Methylcyclopentane for synthesis, Sigma-Aldrich®
Methylcyclopentane
Formula: C₆H₁₂
MW: 84.16 g/mol
MDL Number: MFCD00001382
CAS Number: 96-37-7
UN: 2298
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -142 °C
Molar Mass 84.16 g/mol
Boiling Point 72 °C (1013 mbar)
Vapor Pressure 150 hPa (20 °C)
Flash Point -17 °C
Refractive Index 1.4087 (20 °C, 589 nm)
Density 0.74 g/cm3 (20 °C)
Ignition Point 315 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR