4-Methylcyclohexanone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.20788.0100
8.20788.0500
8.20788.0100
8.20788.0500
EM8.20788.0500CS
2163
USD
CA8.20788.0500
EM8.20788.0500
CA8.20788.0100
EM8.20788.0100
4-Methylcyclohexanone for synthesis, Sigma-Aldrich®
4-Methylcyclohexanone
Formula:
C₇H₁₂O MW: 112.17 g/mol |
MDL Number:
MFCD00001643 CAS Number: 589-92-4 UN: 2297 ADR: 3,III |
Specification Test Results
Solubility | (20 °C) practically insoluble |
Melting Point | -41 °C |
Molar Mass | 112.17 g/mol |
Boiling Point | 169 - 171 °C (1013 hPa) |
Vapor Pressure | 20 hPa (65 °C) |
Flash Point | 48 °C |
Density | 0.92 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...