Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Hexachloroacetone for synthesis

Supplier: MilliporeSigma

Synonyms: Hexachloro-2-propanone

8.20630.0250 8.20630.0250
EM8.20630.0250CS 900.43 USD InStock
EM8.20630.0250 CA8.20630.0250
Hexachloroacetone for synthesis
Hexachloroacetone


Formula: C₃Cl₆O
MW: 264,75 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: -3 °C
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00000796
CAS Number: 116-16-5
EINECS: 204-129-5
UN: 2661
ADR: 6.1,III
Order Now
Specification Test Results
Solubility (20 °C) slightly soluble
Melting Point -3 °C
Molar Mass 264.75 g/mol
Boiling Point 203 - 205 °C
Refractive Index 1.5112 (20 °C, 589 nm)
Density 1.74 g/cm3 (20 °C)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty