Formaldehyde dimethyl acetal for synthesis, Sigma-Aldrich®

Danger

Synonyms: FormalMethylalDimethoxymethane

M-Clarity™ quality level = MQ200

8.20493.0250 8.20493.0250 8.20493.1000 8.20493.1000
EM8.20493.1000CS 875.97 USD
EM8.20493.1000 CA8.20493.0250 CA8.20493.1000 EM8.20493.0250
Formaldehyde dimethyl acetal for synthesis, Sigma-Aldrich®
Formaldehyde dimethyl acetal
Formula: CH₂(OCH₃)₂
MW: 76.1 g/mol
MDL Number: MFCD00008495
CAS Number: 109-87-5
UN: 1234
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/06/92/730692.jpg

Order Now

Specification Test Results

Solubility 285 g/l (20 °C)
Melting Point -105 °C
Molar Mass 76.09 g/mol
Boiling Point 43 °C
Vapor Pressure 440 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.3541 (20 °C, 589 nm)
Explosion Limit 1.6 - 17.6 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 235 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR