3,4-Dichlorophenol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.20450.0100
8.20450.0100
EM8.20450.0100CS
3241
USD
CA8.20450.0100
EM8.20450.0100
3,4-Dichlorophenol for synthesis, Sigma-Aldrich®
3,4-Dichlorophenol
Formula:
C₆H₄Cl₂O MW: 163 g/mol |
MDL Number:
MFCD00002258 CAS Number: 95-77-2 UN: 2020 ADR: 6.1,III |
Specification Test Results
Solubility | 9260 mg/l (25 °C) (experimental) |
Melting Point | 66 - 68 °C |
Molar Mass | 163 g/mol |
Boiling Point | 146 °C (1013 hPa) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...