Sodium-4-hydroxymercuriobenzoate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
4-(Hydroxymercuri)benzoic acid sodium salt
M-Clarity™ quality level = MQ200
8.20308.0005
8.20308.0005
EM8.20308.0005CS
89356
USD
EM8.20308.0005
CA8.20308.0005
Sodium-4-hydroxymercuriobenzoate for synthesis, Sigma-Aldrich®
Sodium 4-hydroxymercuriobenzoate
Formula:
C₇H₅HgNaO₃ |
CAS Number:
138-85-2 UN: 2025 ADR: 6.1,III |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Molar Mass | 360.7 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...