2-Chloro-N,N-dimethylacetamide for synthesis, Sigma-Aldrich®

Warning


M-Clarity™ quality level = MQ200

8.20290.0025 8.20290.0025
EM8.20290.0025CS 12065.35 USD
EM8.20290.0025 CA8.20290.0025
2-Chloro-N,N-dimethylacetamide for synthesis, Sigma-Aldrich®
2-Chloro-N,N-dimethylacetamide
Formula: C₄H₈ClNO
MDL Number: MFCD00039360
CAS Number: 2675-89-0

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 121.56 g/mol
Boiling Point 98 - 100 °C (15 hPa)
Density 1.18 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR