tert-Butylbenzene for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.20240.0500 8.20240.0100 8.20240.0500
EM8.20240.0500CS 670.67 USD
CA8.20240.0500 CA8.20240.0100 EM8.20240.0500
tert-Butylbenzene for synthesis, Sigma-Aldrich®
tert-Butylbenzene
Formula: C₁₀H₁₄
MW: 134.22 g/mol
MDL Number: MFCD00008816
CAS Number: 98-06-6
UN: 2709
ADR: 3,III

Order Now

Specification Test Results

Solubility 29.5 mg/l (25 °C)
Melting Point -58 °C
Molar Mass 134.22 g/mol
Boiling Point 64 °C (27 hPa)
Vapor Pressure 1.33 hPa (13 °C)
Flash Point 44 °C
Refractive Index 1.492 (20 °C, 589 nm)
Explosion Limit 0.8 - 5.6 %(V)
Density 0.87 g/cm3 (20 °C)
Ignition Point 445 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR