(±)-Ethyl-2-bromopropionate for synthesis, Sigma-Aldrich®

Danger

Synonyms: Ethyl 2-bromopropionateDL-Ethyl 2-bromopropionate

M-Clarity™ quality level = MQ200

8.20219.0250 8.20219.0250
EM8.20219.0250CS 1439.57 USD
CA8.20219.0250 EM8.20219.0250
(±)-Ethyl-2-bromopropionate for synthesis, Sigma-Aldrich®
(±)-Ethyl 2-bromopropionate
Formula: C₅H₉BrO₂
MW: 181.03 g/mol
MDL Number: MFCD00000144
CAS Number: 535-11-5
UN: 1992
ADR: 3 (6.1),III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 181.02 g/mol
Boiling Point 156 - 160 °C (1013 hPa)
Flash Point 52 °C
Refractive Index 1.447 (20 °C, 589 nm)
Density 1.39 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR