2-Methylimidazole for synthesis, Sigma-Aldrich®

Danger

Synonyms: 2-Methyl-1H-imidazole

M-Clarity™ quality level = MQ200

8.18964.0005 8.18964.0005 8.18964.0250 8.18964.0250
EM8.18964.0250CS 1596.11 USD
EM8.18964.0250 CA8.18964.0250 EM8.18964.0005 CA8.18964.0005
2-Methylimidazole for synthesis, Sigma-Aldrich®
2-Methylimidazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
MDL Number: MFCD00005190
CAS Number: 693-98-1
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Solubility 780 g/l (20 °C)
Melting Point 144 - 147 °C
Molar Mass 82.11 g/mol
Bulk Density 550 kg/m3
Boiling Point 270 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 155 °C DIN 51758
pH 10.5 - 11.5 (100 g/l, H2O, 20 °C)
Ignition Point 600 °C DIN 51794

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR