2,5-Dichlorobenzoic acid for synthesis, Sigma-Aldrich®

Warning


M-Clarity™ quality level = MQ200

8.18834.0025 8.18834.0025
EM8.18834.0025CS 10566.41 USD
CA8.18834.0025 EM8.18834.0025
2,5-Dichlorobenzoic acid for synthesis, Sigma-Aldrich®
2,5-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
MDL Number: MFCD00002416
CAS Number: 50-79-3

Order Now

Specification Test Results

Solubility 0.8 g/l (14 °C)
Melting Point 151 - 154 °C
Molar Mass 191.01 g/mol
Boiling Point 300 - 302 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR