Allyl acetoacetate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Acetoacetic acid allyl ester
, Allyl Acetoacetate
M-Clarity™ quality level = MQ200
8.18694.0100
8.18694.0100
EM8.18694.0100CS
2265
USD
CA8.18694.0100
EM8.18694.0100
Allyl acetoacetate for synthesis, Sigma-Aldrich®
Allyl acetoacetate
Formula:
C₇H₁₀O₃ MW: 142.1525 g/mol |
MDL Number:
MFCD00009811 CAS Number: 1118-84-9 UN: 2810 ADR: 6.1,III |
Specification Test Results
Solubility | 48 g/l (20 °C) |
Molar Mass | 142.15 g/mol |
Boiling Point | 87 - 91 °C (13 hPa) |
Vapor Pressure | 0.2 hPa (20 °C) |
Flash Point | 67 °C |
Refractive Index | 1.438 (25 °C, 589 nm) |
Explosion Limit | 1.15 %(V) |
Density | 1.038 g/cm3 (20 °C) |
pH | 3.7 (48 g/l, H2O, 20 °C) |
Ignition Point | 300 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...