Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2-Methoxyphenyl)acetic acid for synthesis

Supplier: MilliporeSigma


8.18671.0025 8.18671.0025
EM8.18671.0025CS 7876.96 USD InStock
EM8.18671.0025 CA8.18671.0025
(2-Methoxyphenyl)acetic acid for synthesis
(2-Methoxyphenyl)acetic acid


Formula: (C₆H₄OCH₃)CH₂COOH
MW: 166,18 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 120 to 123 °C
Storage Temperature: ambient
MDL Number: MFCD00004321
CAS Number: 93-25-4
EINECS: 202-231-4
Order Now
Specification Test Results
Solubility 9.2 g/l (20 °C)
Melting Point 120 - 123 °C
Molar Mass 166.18 g/mol


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty