Butylmethylamine for synthesis, Sigma-Aldrich®

Danger

Synonyms: N-MethylbutylamineN-Butylmethylamine

M-Clarity™ quality level = MQ200

8.18639.0500 8.18639.0500 8.18639.0100 8.18639.0100
EM8.18639.0100CS 1352.74 USD
EM8.18639.0100 CA8.18639.0100 EM8.18639.0500 CA8.18639.0500
Butylmethylamine for synthesis, Sigma-Aldrich®
Butylmethylamine
Formula: C₅H₁₃N
MW: 87.16 g/mol
MDL Number: MFCD00009426
CAS Number: 110-68-9
UN: 2945
ADR: 3 (8),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -75 °C
Molar Mass 87.17 g/mol
Boiling Point 90 - 92 °C (1013 hPa)
Flash Point -4 °C
Refractive Index 1.4011 (20 °C, 589 nm)
Explosion Limit 1.3 %(V)
Density 0.74 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR