Dihydrocoumarin for synthesis

Supplier: MilliporeSigma

Synonyms: 1,2-Benzodihydropyrone

8.18454.0100 8.18454.0100
EM8.18454.0100CS 1171.51 USD
CA8.18454.0100 EM8.18454.0100
Dihydrocoumarin for synthesis
Dihydrocoumarin
Formula: C₉H₈O₂
MDL Number: MFCD00006881
CAS Number: 119-84-6

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 19 °C
Molar Mass 148.16 g/mol
Boiling Point 272 °C (1013 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 100 °C
Refractive Index 1.5528 (25 °C, 589 nm)
Density 1.19 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR