9-Aminoacridine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
9-AA
M-Clarity™ quality level = MQ200
8.18362.0010
8.18362.0010
EM8.18362.0010CS
63327
USD
CA8.18362.0010
EM8.18362.0010
9-Aminoacridine for synthesis, Sigma-Aldrich®
9-Aminoacridine
Specification Test Results
Solubility | (20 °C) practically insoluble |
Melting Point | 231 - 234 °C |
Molar Mass | 194.23 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...