Pentafluorophenol for synthesis, Sigma-Aldrich®

Warning

Synonyms: 2,3,4,5,6-Pentafluorophenol

M-Clarity™ quality level = MQ200

8.18100.0025 8.18100.0005 8.18100.0005 8.18100.0025 8.18100.0100 8.18100.0100
EM8.18100.0005CS 43832.65 USD
EM8.18100.0005 EM8.18100.0025 EM8.18100.0100 CA8.18100.0005 CA8.18100.0025 CA8.18100.0100
Pentafluorophenol for synthesis, Sigma-Aldrich®
Pentafluorophenol
Formula: C₆HF₅O
MW: 184.07 g/mol
MDL Number: MFCD00002156
CAS Number: 771-61-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 34 - 36 °C
Molar Mass 184.07 g/mol
Boiling Point 143 °C
Flash Point 72 °C
Refractive Index 1.427 (20 °C, 589 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR