Aluminium triethanolate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Aluminumethoxide
, Aluminum ethoxide
, Aluminium triethylate
M-Clarity™ quality level = MQ200
8.18097.0050
8.18097.0050
EM8.18097.0050CS
9120.35
USD
EM8.18097.0050
CA8.18097.0050
Aluminium triethanolate for synthesis, Sigma-Aldrich®
Aluminium triethanolate
Formula:
C₆H₁₅AlO₃ MW: 162.16 g/mol |
MDL Number:
MFCD00009658 CAS Number: 555-75-9 UN: 1325 ADR: 4.1,II |
Specification Test Results
Solubility | (20 °C) (rigorous decomposition) |
Melting Point | 128 - 131 °C |
Molar Mass | 162.16 g/mol |
Boiling Point | 319 - 321 °C |
Density | 1.142 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...