Formic acid-triethylamine 5:2 addition compound for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: TEAFFormic acid triethylamine complex 5:2Formic acid - triethylamine (5:2) Azeotrope

M-Clarity™ quality level = MQ200

8.18028.0050 8.18028.0050
EM8.18028.0050CS 15144 USD
EM8.18028.0050 CA8.18028.0050
Formic acid-triethylamine 5:2 addition compound for synthesis, Sigma-Aldrich®
Formic acid-triethylamine 5:2 addition compound
Formula: C₁₇H₄₀N₂O₁₀
MW: 432.51 g/mol
MDL Number: MFCD00040600
CAS Number: 15077-13-1
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 432.51 g/mol
Boiling Point 192 - 194 °C
Flash Point 110 °C
Density 1.03 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR