Triclosan for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
5-Chloro-2-(2, 4-dichlorophenoxy) phenol
, 5-Chloro-2-(2,4-dichlorophenoxy)phenol
M-Clarity™ quality level = MQ200
8.16100.0025
8.16100.0025
EM8.16100.0025CS
10895
USD
CA8.16100.0025
EM8.16100.0025
Triclosan for synthesis, Sigma-Aldrich®
Triclosan
Formula:
C₁₂H₇Cl₃O₂ MW: 289.54 g/mol |
MDL Number:
MFCD00800992 CAS Number: 3380-34-5 UN: 3077 ADR: 9,III |
Specification Test Results
Solubility | 0.01 g/l (20 °C) |
Melting Point | 55 - 60 °C |
Molar Mass | 289.55 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...