2,5-Diethyl-1,3,4-thiadiazole for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.15100.0025
8.15100.0025
EM8.15100.0025CS
14528
USD
CA8.15100.0025
EM8.15100.0025
2,5-Diethyl-1,3,4-thiadiazole for synthesis, Sigma-Aldrich®
2,5-Diethyl-1,3,4-thiadiazole
Specification Test Results
Solubility | (20 °C) slightly soluble |
Molar Mass | 142.22 g/mol |
Boiling Point | 105 °C (18.7 hPa) |
Density | 1.071 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...