Isobornyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14998.0250 8.14998.0250
EM8.14998.0250CS 575 USD
CA8.14998.0250 EM8.14998.0250
Isobornyl acetate for synthesis, Sigma-Aldrich®
Isobornyl acetate
Formula: C₁₂H₂₀O₂
MDL Number: MFCD00135943
CAS Number: 125-12-2

Order Now

Specification Test Results

Solubility 0.16 g/l (20 °C)
Molar Mass 196.29 g/mol
Boiling Point 215 °C (1013 hPa)
Vapor Pressure 0.13 hPa (20 °C)
Flash Point 88 °C
Density 0.98 g/cm3 (20 °C)
Ignition Point 430 °C DIN 51794

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR