Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

5-Bromo-2-chlorobenzoic acid for synthesis


8.14989.0101 8.14989.0101
EM8.14989.0101CS 9379.1 USD InStock
CA8.14989.0101 EM8.14989.0101
5-Bromo-2-chlorobenzoic acid for synthesis
5-Bromo-2-chlorobenzoic acid


Formula: C₇H₄BrClO₂
MW: 235,46 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 157 to 160 °C
Storage Temperature: ambient
MDL Number: MFCD00002415
CAS Number: 21739-92-4
EINECS: 244-558-5
Order Now
Specification Test Results
Solubility 2.63 g/l (20 °C)
Melting Point 157 - 160 °C
Molar Mass 235.47 g/mol

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty