2-(2-Chloroethyl)-1-methylpiperidinium chloride for synthesis, Sigma-Aldrich®

Warning

Synonyms: 2-(2-Chloroethyl)-1-methylpiperidine hydrochloride

M-Clarity™ quality level = MQ200

8.14738.0010 8.14738.0010
EM8.14738.0010CS 15366.65 USD
EM8.14738.0010 CA8.14738.0010
2-(2-Chloroethyl)-1-methylpiperidinium chloride for synthesis, Sigma-Aldrich®
2-(2-Chloroethyl)-1-methylpiperidinium chloride
Formula: C₈H₁₇Cl₂N
MDL Number: MFCD00039047
CAS Number: 58878-37-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 133 - 137 °C
Molar Mass 198.14 g/mol
pH 5.9 (H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR