Potassium 2,5-dihydroxybenzenesulfonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,5-Dihydroxybenzenesulphonic acid potassium salPotassium 2,5-dihydroxybenzenesulfonateHydroquinonesulphonic acid potassium salt2,5-Dihydroxybenzenesulfonic acid potassium salPotassium 2,5-dihydroxybenzenesulphonateHydroquinonesulfonic acid potassium salt

M-Clarity™ quality level = MQ200

8.14729.0250 8.14729.0250
EM8.14729.0250CS 2561 USD
CA8.14729.0250 EM8.14729.0250
Potassium 2,5-dihydroxybenzenesulfonate for synthesis, Sigma-Aldrich®
Potassium 2,5-dihydroxybenzenesulphonate
Formula: C₆H₅KO₅S
MW: 228.27 g/mol
MDL Number: MFCD00007475
CAS Number: 21799-87-1

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 250 - 251 °C
Molar Mass 228.27 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR