4-Amino-2,2,6,6-tetramethylpiperidine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.14722.0100
8.14722.0100
EM8.14722.0100CS
1530
USD
EM8.14722.0100
CA8.14722.0100
4-Amino-2,2,6,6-tetramethylpiperidine for synthesis, Sigma-Aldrich®
4-Amino-2,2,6,6-tetramethylpiperidine
Formula:
C₉H₂₀N₂ |
MDL Number:
MFCD00005984 CAS Number: 36768-62-4 UN: 2735 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 12 - 15 °C |
Molar Mass | 156.27 g/mol |
Boiling Point | 199 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 75 °C |
Refractive Index | 1.469 (25 °C) |
Density | 0.893 g/cm3 (20 °C) |
pH | 12.3 (99 g/l, H2O, 20 °C) |
Ignition Point | 300 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...