Dimethoxy(diisopropyl)silane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Diisopropyldimethoxysilane

M-Clarity™ quality level = MQ200

8.14719.0025 8.14719.0025
EM8.14719.0025CS 12757.56 USD
CA8.14719.0025 EM8.14719.0025
Dimethoxy(diisopropyl)silane for synthesis, Sigma-Aldrich®
Dimethoxy(diisopropyl)silane
Formula: C₈H₂₀O₂Si
MW: 176.33 g/mol
MDL Number: MFCD00239343
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Molar Mass 176.33 g/mol
Boiling Point 164 °C (1013 hPa)
Vapor Pressure 2.14 hPa (20 °C)
Flash Point 44 °C
Density 0.88 g/cm3 (20 °C)
Ignition Point 240 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR