2,2,6,6-Tetramethylpiperidinooxy (TEMPO) for synthesis, Sigma-Aldrich®

Danger

Synonyms: 2,2,6,6-Tetramethylpiperidine 1-oxyl2,2,6,6-Tetramethyl-1-oxylpiperidineTEMPO

M-Clarity™ quality level = MQ200

8.14681.0005 8.14681.0025 8.14681.0025 8.14681.0005
EM8.14681.0025CS 18885.3 USD
EM8.14681.0025 EM8.14681.0005 CA8.14681.0005 CA8.14681.0025
2,2,6,6-Tetramethylpiperidinooxy (TEMPO) for synthesis, Sigma-Aldrich®
2,2,6,6-Tetramethylpiperidinooxy (TEMPO, free radical)
Formula: C₉H₁₈NO
MW: 156.25 g/mol
MDL Number: MFCD00009599
CAS Number: 2564-83-2
UN: 3263
ADR: 8,II

Order Now

Specification Test Results

Solubility 9.7 g/l (20 °C)
Melting Point 33 - 38 °C
Molar Mass 156.25 g/mol
Vapor Pressure 0.4 hPa (20 °C)
Flash Point 67 °C
Density 0.912 g/cm3 (40 °C)
pH 8.3 (9 g/l, H2O, 20 °C)
Ignition Point 275 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR