2-Bromoethyl ethyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.14667.0025 8.14667.0025
EM8.14667.0025CS 14067.09 USD
CA8.14667.0025 EM8.14667.0025
2-Bromoethyl ethyl ether for synthesis, Sigma-Aldrich®
2-Bromoethyl ethyl ether
Formula: C₄H₉BrO
MDL Number: MFCD00000237
CAS Number: 592-55-2
UN: 2340
ADR: 3,II

Order Now

Specification Test Results

Molar Mass 153.01 g/mol
Boiling Point 125 - 127 °C (1013 hPa)
Flash Point 21 °C
Density 1.35 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR