Diisopropyl azodicarboxylate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Diisopropyl diazene-1,2-dicarboxylate

M-Clarity™ quality level = MQ200

8.14566.0051 8.14566.0051
EM8.14566.0051CS 11271 USD
CA8.14566.0051 EM8.14566.0051
Diisopropyl azodicarboxylate for synthesis, Sigma-Aldrich®
Diisopropyl azodicarboxylate
Formula: C₈H₁₄N₂O₄
MW: 202.21 g/mol
MDL Number: MFCD00008875
CAS Number: 2446-83-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 202.2 g/mol
Boiling Point 75 °C (33.3 Pa)
Vapor Pressure 1.04 hPa (20 °C)
Flash Point 106 °C
Density 1.03 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR