Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-3-(Methylamino)-1,2-propanediol for synthesis

Synonyms: (±)-3-(Methylamino)-1,2-dihydroxy propane

8.14507.0025 8.14507.0025
EM8.14507.0025CS 4674.85 USD InStock
CA8.14507.0025 EM8.14507.0025
(±)-3-(Methylamino)-1,2-propanediol for synthesis
(±)-3-(Methylamino)-1,2-propanediol


Formula: C₄H₁₁NO₂
MW: 105,14 g/mol
MDL Number: MFCD00082732
CAS Number: 40137-22-2
EINECS: 254-809-0
Order Now
Specification Test Results
Solubility (20 °C) soluble
Molar Mass 105.14 g/mol
Boiling Point 139 - 241 °C (995 hPa)
Flash Point 152 °C
Density 1.09 g/cm3 (20 °C)
pH 11.5 (100 g/l, H2O, 20 °C)

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty