Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Bromomethyl)cyclobutane for synthesis

Synonyms: Cyclobutyl methyl bromide

8.14484.0001 8.14484.0001
EM8.14484.0001CS 4716.55 USD InStock
CA8.14484.0001 EM8.14484.0001
(Bromomethyl)cyclobutane for synthesis
(Bromomethyl)cyclobutane


Formula: C₅H₉Br
MW: 149,03 g/mol
Boiling Pt: 123 to 124 °C (1013 hPa)
Density: 1.35 g/cm³ (20 °C)
Flash Pt: 41 °C
Storage Temperature: ambient
MDL Number: MFCD00040960
CAS Number: 17247-58-4
EINECS: 000-000-0
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Solubility (20 °C) (decomposition)
Molar Mass 149.03 g/mol
Boiling Point 123 - 124 °C
Flash Point 41 °C
Refractive Index 1.4726 (20 °C, 589 nm)
Density 1.35 g/cm3 (20 °C)

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty