Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,3,5-Trimethylcyclohexanol cis- and trans- mixture for synthesis

Supplier: MilliporeSigma


8.14442.0005 8.14442.0005 8.14442.0250 8.14442.0250
EM8.14442.0005CS 1954.29 USD InStock
CA8.14442.0005 EM8.14442.0005 EM8.14442.0250 CA8.14442.0250
3,3,5-Trimethylcyclohexanol cis- and trans- mixture for synthesis
3,3,5-Trimethylcyclohexanol


Formula: C₉H₁₈O
MW: 142,24 g/mol
Boiling Pt: 193 °C (1013 hPa)
Melting Pt: 30 to 32 °C
Density: 0.9006 g/cm³ (16 °C)
Storage Temperature: ambient
CAS Number: 116-02-9
EINECS: 204-122-7
Order Now
Specification Test Results
Solubility 1.8 g/l (20 °C)
Melting Point 30 - 32 °C
Molar Mass 142.24 g/mol
Boiling Point 193 - 196 °C (1013 hPa)
Vapor Pressure 1 hPa (50 °C)
Flash Point 81 °C DIN 51758
Refractive Index 1.439 (60 °C, 589 nm)
Explosion Limit 1.5 %(V)
Density 0.86 g/cm3 (60 °C)
pH 7 (1.8 g/l, H2O, 20 °C)
Ignition Point 375 °C DIN 51794


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty