Octyltriethoxysilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Triethoxy(octyl)silane

M-Clarity™ quality level = MQ200

8.14414.0100 8.14414.0100
EM8.14414.0100CS 896 USD
EM8.14414.0100 CA8.14414.0100
Octyltriethoxysilane for synthesis, Sigma-Aldrich®
Octyltriethoxysilane
Formula: C₁₄H₃₂O₃Si
MW: 276.49 g/mol
MDL Number: MFCD00039883
CAS Number: 2943-75-1

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 276.49 g/mol
Boiling Point 98 - 99 °C (2.7 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 109 °C
Refractive Index 1.415 (25 °C, 589 nm)
Density 0.87 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR