2-Mercaptoethanesulfonic acid sodium salt (mesna) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Sodium 2-mercaptoethanesulfonateSodium 2-mercaptoethanesulphonate

M-Clarity™ quality level = MQ200

8.10595.0010 8.10595.0010
EM8.10595.0010CS 10019 USD
EM8.10595.0010 CA8.10595.0010
2-Mercaptoethanesulfonic acid sodium salt (mesna) for synthesis, Sigma-Aldrich®
2-Mercaptoethanesulphonic acid sodium salt (Mesna)
Formula: C₂H₅NaO₃S₂
MW: 164.18 g/mol
MDL Number: MFCD00007535
CAS Number: 19767-45-4

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 164.18 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR