2-Picoline for synthesis, Sigma-Aldrich®

Warning

Synonyms: 2-Methylpyridineα-Picoline

M-Clarity™ quality level = MQ200

8.09722.0500 8.09722.1000 8.09722.1000 8.09722.0100
EM8.09722.1000CS 503.11 USD
CA8.09722.1000 CA8.09722.0500 CA8.09722.0100 EM8.09722.1000
2-Picoline for synthesis, Sigma-Aldrich®
2-Picoline
Formula: C₆H₇N
MW: 93.13 g/mol
MDL Number: MFCD00006332
CAS Number: 109-06-8
UN: 2313
ADR: 3,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -70 °C
Molar Mass 93.13 g/mol
Boiling Point 129 °C (1013 hPa)
Vapor Pressure 12 hPa (20 °C)
Flash Point 27 °C
Refractive Index 1.501 (20 °C, 589 nm)
Explosion Limit 1.4 - 8.6 %(V)
Density 0.94 g/cm3 (20 °C)
pH 8.5 (100 g/l, H2O, 20 °C)
Ignition Point 535 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR