Tetramethyltin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Tin tetramethylTetramethylstannane

M-Clarity™ quality level = MQ200

8.08706.0010 8.08706.0010
EM8.08706.0010CS 197014 USD
EM8.08706.0010 CA8.08706.0010
Tetramethyltin for synthesis, Sigma-Aldrich®
Tetramethyltin
Formula: C₄H₁₂Sn
MW: 178.85 g/mol
MDL Number: MFCD00008278
CAS Number: 594-27-4
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point -54 °C
Molar Mass 178.82 g/mol
Boiling Point 74 - 75 °C (1013 hPa)
Flash Point -12 °C
Refractive Index 1.441 (20 °C, 589 nm)
Density 1.29 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR