Fentin chloride for synthesis, Sigma-Aldrich®

Danger

Synonyms: ChlorotriphenylstannaneTriphenyltin chloride

M-Clarity™ quality level = MQ200

8.08395.0100
EM8.08395.0100CS 1549.45 USD
EM8.08395.0100
Fentin chloride for synthesis, Sigma-Aldrich®
Fentin chloride
Formula: C₁₈H₁₅ClSn
MW: 385.48 g/mol
MDL Number: MFCD00000519
CAS Number: 639-58-7
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 0.04 g/l (20 °C)
Melting Point 102 - 105 °C
Molar Mass 385.45 g/mol
Boiling Point 240 °C (18 hPa) (decomposition)
Vapor Pressure  - 0.02 hPa (20 °C)
Flash Point 150 °C
Density 1.49 g/cm3 (20 °C)
Ignition Point 250 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR