3-Sulfolene for synthesis, Sigma-Aldrich®

Warning

Synonyms: 3-sulfoleneButadiene sulphoneButadiene sulfone3-sulpholene

M-Clarity™ quality level = MQ200

8.07994.0250 8.07994.0250
EM8.07994.0250CS 1117.85 USD
EM8.07994.0250 CA8.07994.0250
3-Sulfolene for synthesis, Sigma-Aldrich®
3-Sulpholene
Formula: C₄H₆O₂S
MW: 118.16 g/mol
MDL Number: MFCD00005481
CAS Number: 77-79-2

Order Now

Specification Test Results

Solubility 130 g/l (20 °C)
Melting Point 62 - 65 °C
Molar Mass 118.16 g/mol
Bulk Density 840 kg/m3
Flash Point 112 °C
Density 1.31 g/cm3 (15.6 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR