Phenylhydrazine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.07250.1000 8.07250.0005 8.07250.1000 8.07250.0005 8.07250.0250 8.07250.0250
EM8.07250.0005CS 1896 USD
EM8.07250.0005 CA8.07250.1000 CA8.07250.0250 EM8.07250.0250 CA8.07250.0005 EM8.07250.1000
Phenylhydrazine for synthesis, Sigma-Aldrich®
Phenylhydrazine
Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00007573
CAS Number: 100-63-0
UN: 2572
ADR: 6.1,II

Order Now

Specification Test Results

Solubility 145 g/l (25 °C)
Melting Point 19.6 °C
Molar Mass 108.14 g/mol
Boiling Point 244 °C (1013 hPa)
Vapor Pressure 0.1 hPa (20 °C)
Flash Point 88 °C
Refractive Index 1.607 (20 °C, 589 nm)
Explosion Limit 1.1 %(V)
Density 1.10 g/cm3 (20 °C)
pH (H2O, 25 °C) alkaline,(undiluted)
Ignition Point 195 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR