Sodium 3-nitrobenzenesulfonate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Sodium 3-nitrobenzenesulphonate
, 3-Nitrobenzenesulfonic acid, sodium salt
, 3-Nitrobenzenesulphonic acid, sodium salt
M-Clarity™ quality level = MQ200
8.06548.0100
8.06548.0100
8.06548.1000
8.06548.1000
EM8.06548.1000CS
612
USD
EM8.06548.1000
EM8.06548.0100
CA8.06548.1000
CA8.06548.0100
Sodium 3-nitrobenzenesulfonate for synthesis, Sigma-Aldrich®
Sodium 3-nitrobenzenesulphonate
Formula:
C₆H₄NNaO₅S MW: 225.16 g/mol |
MDL Number:
MFCD00007490 CAS Number: 127-68-4 |
Specification Test Results
Solubility | 200 g/l (20 °C) |
Melting Point | 350 °C (decomposition) |
Molar Mass | 225.16 g/mol |
Bulk Density | 450 kg/m3 |
pH | 8 (50 g/l, H2O, 23 °C) |
Ignition Point | 355 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...