N-Methylaniline for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.06096.1000 8.06096.0100 8.06096.0100 8.06096.1000
EM8.06096.1000CS 629.7 USD
CA8.06096.1000 CA8.06096.0100 EM8.06096.1000 EM8.06096.0100
N-Methylaniline for synthesis, Sigma-Aldrich®
N-Methylaniline
Formula: C₇H₉N
MW: 107.16 g/mol
MDL Number: MFCD00008283
CAS Number: 100-61-8
UN: 2294
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 5.62 g/l (25 °C)
Melting Point -57 °C
Molar Mass 107.16 g/mol
Boiling Point 196 °C (1013 hPa)
Vapor Pressure 0.5 hPa (20 °C)
Flash Point 86 °C
Refractive Index 1.5704 (20 °C, 589 nm)
Density 0.99 g/cm3 (20 °C)
pH 7.6 (1 g/l, H2O, 20 °C)
Ignition Point 500 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR